Assay Details
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266073.1 | NP_001253002.1 | 455 | 95 | ||
XM_028850352.1 | XP_028706185.1 | 519 | 95 | |||
XM_028850353.1 | XP_028706186.1 | 519 | 95 | |||
XM_028850354.1 | XP_028706187.1 | 538 | 95 | |||
XM_028850355.1 | XP_028706188.1 | 538 | 95 | |||
XM_028850358.1 | XP_028706191.1 | 301 | 95 | |||
XM_028850359.1 | XP_028706192.1 | 813 | 95 | |||
XM_028850360.1 | XP_028706193.1 | 193 | 95 | |||
XM_028850362.1 | XP_028706195.1 | 301 | 95 | |||
GenBank mRNA | JU319594.1 | - | 463 | 95 | ||
JU319595.1 | - | 463 | 95 | |||
JU471722.1 | - | 455 | 95 | |||
JU471723.1 | - | 455 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028850358.1, XM_028850359.1, XM_028850360.1, XM_028850362.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
