Assay Details
| Gene Symbol: HUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088000.4 | XP_001088000.1 | 861 | 76 | ||
| XM_015133577.2 | XP_014989063.1 | 1207 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HUS1| Gene Symbol: HUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088000.4 | XP_001088000.1 | 861 | 76 | ||
| XM_015133577.2 | XP_014989063.1 | 1207 | 76 | |||
Set Membership: |
|