Assay Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194573.2 | NP_001181502.1 | 587 | 114 | ||
| XM_015113654.2 | XP_014969140.1 | 722 | 114 | |||
| XM_028832776.1 | XP_028688609.1 | 393 | 114 | |||
More Information
Set Membership: |
|

OVOL1
Genomic Map