Assay Details
| Gene Symbol: PNPLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193844.2 | NP_001180773.1 | 588 | 139 | ||
| XM_015126905.2 | XP_014982391.1 | 583 | 139 | |||
| XM_028841796.1 | XP_028697629.1 | 583 | 139 | |||
| XM_028841797.1 | XP_028697630.1 | 583 | 139 | |||
| XM_028841798.1 | XP_028697631.1 | 583 | 139 | |||
| XM_028841799.1 | XP_028697632.1 | 583 | 139 | |||
| GenBank mRNA | JU323916.1 | - | 505 | 139 | ||
| JV662738.1 | - | 355 | 139 | |||
More Information
Set Membership: |
|

PNPLA4
Genomic Map