Assay Details
| Gene Symbol: RAB11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261789.1 | NP_001248718.1 | 554 | 142 | ||
| GenBank mRNA | JU336914.1 | - | 549 | 142 | ||
| JU477650.1 | - | 472 | 142 | |||
| JV048581.1 | - | 554 | 142 | |||
| JV664470.1 | - | 557 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB11A
Genomic Map