Assay Details
| Gene Symbol: KIF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266387.1 | NP_001253316.1 | 1924 | 88 | ||
| XM_028827342.1 | XP_028683175.1 | 2044 | 88 | |||
| GenBank mRNA | JU328612.1 | - | 661 | 88 | ||
| JU471068.1 | - | 1924 | 88 | |||
More Information
Set Membership: |
|

KIF3B
Genomic Map