Assay Details
| Gene Symbol: POLR2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267787.1 | NP_001254716.1 | 266 | 97 | ||
| GenBank mRNA | JU332203.1 | - | 285 | 97 | ||
| JU471587.1 | - | 266 | 97 | |||
| JV045438.1 | - | 281 | 97 | |||
| JV619748.1 | - | 305 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLR2D
Genomic Map