Assay Details
| Gene Symbol: CDYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847858.1 | XP_028703691.1 | 1478 | 152 | ||
| XM_028847859.1 | XP_028703692.1 | 1499 | 152 | |||
| XM_028847860.1 | XP_028703693.1 | 1478 | 152 | |||
| XM_028847861.1 | XP_028703694.1 | 1164 | 152 | |||
| GenBank mRNA | JU337175.1 | - | 1163 | 152 | ||
| JU496588.1 | - | 1111 | 152 | |||
| JV048748.1 | - | 1286 | 152 | |||
More Information
Set Membership: |
|

CDYL
Genomic Map