Assay Details
| Gene Symbol: CHST10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257866.1 | NP_001244795.1 | 330 | 81 | ||
| XM_015112950.2 | XP_014968436.1 | 334 | 81 | |||
| XM_015112952.2 | XP_014968438.1 | 841 | 81 | |||
| XM_028831569.1 | XP_028687402.1 | 796 | 81 | |||
| XM_028831570.1 | XP_028687403.1 | 796 | 81 | |||
| XM_028831571.1 | XP_028687404.1 | 796 | 81 | |||
| GenBank mRNA | JU321483.1 | - | 330 | 81 | ||
| JU472736.1 | - | 272 | 81 | |||
| JV048764.1 | - | 195 | 81 | |||
More Information
Set Membership: |
|

CHST10
Genomic Map