Assay Details
| Gene Symbol: TP53I3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266424.1 | NP_001253353.1 | 968 | 102 | ||
| XM_015111808.2 | XP_014967294.1 | 1103 | 102 | |||
| XM_028831476.1 | XP_028687309.1 | 1103 | 102 | |||
| GenBank mRNA | JU477931.1 | - | 968 | 102 | ||
More Information
Set Membership: |
|

TP53I3
Genomic Map