Assay Details
| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258043.1 | NP_001244972.1 | 767 | 91 | ||
| GenBank mRNA | JU470662.1 | - | 767 | 91 | ||
| JU470663.1 | - | 767 | 91 | |||
| JU470664.1 | - | 767 | 91 | |||
More Information
Set Membership: |
|

AQP3
Genomic Map