Assay Details
| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082881.4 | XP_001082881.3 | 1426 | 123 | ||
| XM_015116867.2 | XP_014972353.2 | 1205 | 123 | |||
| XM_015116869.2 | XP_014972355.2 | 1048 | 123 | |||
| XM_028834870.1 | XP_028690703.1 | 1437 | 123 | |||
| XM_028834871.1 | XP_028690704.1 | 1239 | 123 | |||
| GenBank mRNA | JV043595.1 | - | 1243 | 123 | ||
| JV638745.1 | - | 1331 | 123 | |||
More Information
Set Membership: |
|

NR5A1
Genomic Map