Assay Details
| Gene Symbol: MECP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257542.1 | NP_001244471.1 | 540 | 89 | ||
| XM_015128480.2 | XP_014983966.1 | 513 | 89 | |||
| XM_015128481.2 | XP_014983967.1 | 2636 | 89 | |||
| XM_028841735.1 | XP_028697568.1 | 2748 | 89 | |||
| GenBank mRNA | JU334548.1 | - | 540 | 89 | ||
| JU472107.1 | - | 509 | 89 | |||
| JV047084.1 | - | 509 | 89 | |||
| JV636161.1 | - | 451 | 89 | |||
| JV651726.1 | - | 575 | 89 | |||
More Information
Set Membership: |
|

MECP2
Genomic Map