Assay Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141249.2 | XP_014996735.1 | 1596 | 143 | ||
| XM_015141263.2 | XP_014996749.1 | 3886 | 143 | |||
More Information
Set Membership: |
|

ROR1
Genomic Map