Assay Details
| Gene Symbol: SSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261346.1 | NP_001248275.1 | 358 | 78 | ||
| GenBank mRNA | JU321370.1 | - | 365 | 78 | ||
| JV043382.1 | - | 358 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSPN| Gene Symbol: SSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261346.1 | NP_001248275.1 | 358 | 78 | ||
| GenBank mRNA | JU321370.1 | - | 365 | 78 | ||
| JV043382.1 | - | 358 | 78 | |||