Assay Details
| Gene Symbol: GOLGA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257502.1 | NP_001244431.1 | 1672 | 105 | ||
| XM_028850242.1 | XP_028706075.1 | 1628 | 105 | |||
| GenBank mRNA | JU323005.1 | - | 1672 | 105 | ||
| JU478099.1 | - | 1636 | 105 | |||
| JU478100.1 | - | 1610 | 105 | |||
| JV044264.1 | - | 1633 | 105 | |||
More Information
Set Membership: |
|

GOLGA5
Genomic Map