Assay Details
| Gene Symbol: HAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098841.4 | XP_001098841.1 | 652 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001098841.4 transcripts.
Set Membership: |
|

HAS2
Genomic Map