Assay Details
| Gene Symbol: LOC702090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128384.2 | XP_014983870.2 | 1863 | 87 | ||
| Gene Symbol: LOC114669842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842109.1 | XP_028697942.1 | 288 | 87 | ||
| XM_028842110.1 | XP_028697943.1 | 249 | 87 | |||
| GenBank mRNA | JV061198.1 | - | 175 | 87 | ||
| JV651576.1 | - | 155 | 87 | |||
More Information
Set Membership: |
|

LOC702090
Genomic Map