Assay Details
| Gene Symbol: RND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261140.1 | NP_001248069.1 | 221 | 72 | ||
| GenBank mRNA | JU336535.1 | - | 221 | 72 | ||
| JV594865.1 | - | 249 | 72 | |||
More Information
Set Membership: |
|

RND2
Genomic Map