Assay Details
| Gene Symbol: BCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101021.4 | XP_001101021.3 | 516 | 72 | ||
| XM_015151294.2 | XP_015006780.1 | 822 | 72 | |||
| XM_015151295.2 | XP_015006781.1 | 747 | 72 | |||
| GenBank mRNA | JU476620.1 | - | 491 | 72 | ||
| JU476621.1 | - | 491 | 72 | |||
| JV720253.1 | - | 239 | 72 | |||
More Information
Set Membership: |
|

BCAT1
Genomic Map