Assay Details
| Gene Symbol: TAF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261556.1 | NP_001248485.1 | 354 | 147 | ||
| XM_028846872.1 | XP_028702705.1 | 447 | 147 | |||
| GenBank mRNA | JU336109.1 | - | 405 | 147 | ||
| JU473713.1 | - | 404 | 147 | |||
| JV048064.1 | - | 354 | 147 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TAF11
Genomic Map