Assay Details
| Gene Symbol: GIPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123140.2 | XP_014978626.1 | 734 | 81 | ||
| XM_028839859.1 | XP_028695692.1 | 955 | 81 | |||
| XM_028839860.1 | XP_028695693.1 | 852 | 81 | |||
| GenBank mRNA | JU319925.1 | - | 857 | 81 | ||
| JU473599.1 | - | 851 | 81 | |||
More Information
Set Membership: |
|

GIPC1
Genomic Map