Assay Details
| Gene Symbol: ACTR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257775.1 | NP_001244704.1 | 721 | 103 | ||
| GenBank mRNA | JU332411.1 | - | 721 | 103 | ||
| JV045841.1 | - | 727 | 103 | |||
| JV670020.1 | - | 708 | 103 | |||
More Information
Set Membership: |
|

ACTR1A
Genomic Map