Assay Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257625.1 | NP_001244554.1 | 139 | 107 | ||
| GenBank mRNA | JU332431.1 | - | 139 | 107 | ||
| JU473893.1 | - | 65 | 107 | |||
| JV045853.1 | - | 91 | 107 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATP6AP2
Genomic Map