Assay Details
| Gene Symbol: SLC23A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140994.2 | XP_014996480.1 | 1023 | 108 | ||
| XM_015140996.2 | XP_014996482.1 | 1023 | 108 | |||
| XM_015140997.2 | XP_014996483.1 | 878 | 108 | |||
More Information
Set Membership: |
|

SLC23A1
Genomic Map