Assay Details
| Gene Symbol: ZMPSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257446.1 | NP_001244375.1 | 1187 | 95 | ||
| GenBank mRNA | JU321800.1 | - | 1187 | 95 | ||
| JU473933.1 | - | 1108 | 95 | |||
| JV043468.1 | - | 1260 | 95 | |||
More Information
Set Membership: |
|

ZMPSTE24
Genomic Map