Assay Details
| Gene Symbol: TSNAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261354.1 | NP_001248283.1 | 524 | 111 | ||
| GenBank mRNA | JU336181.1 | - | 517 | 111 | ||
| JU336182.1 | - | 517 | 111 | |||
| JU336183.1 | - | 517 | 111 | |||
| JU336184.1 | - | 517 | 111 | |||
| JU336185.1 | - | 517 | 111 | |||
| JU336186.1 | - | 517 | 111 | |||
| JU336187.1 | - | 517 | 111 | |||
| JU477036.1 | - | 524 | 111 | |||
| JV048129.1 | - | 524 | 111 | |||
| JV567748.1 | - | 517 | 111 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TSNAX
Genomic Map