Assay Details
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265927.1 | NP_001252856.1 | 591 | 72 | ||
GenBank mRNA | JU320694.1 | - | 604 | 72 | ||
JU472808.1 | - | 580 | 72 | |||
JV045890.1 | - | 591 | 72 |
