Assay Details
| Gene Symbol: NFATC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088054.4 | XP_001088054.2 | 2023 | 95 | ||
| XM_001088166.4 | XP_001088166.2 | 2023 | 95 | |||
| XM_015122239.2 | XP_014977725.1 | 1015 | 95 | |||
| XM_028838228.1 | XP_028694061.1 | 2416 | 95 | |||
| XM_028838229.1 | XP_028694062.1 | 2416 | 95 | |||
| XM_028838230.1 | XP_028694063.1 | 2410 | 95 | |||
| XM_028838231.1 | XP_028694064.1 | 2416 | 95 | |||
More Information
Set Membership: |
|

NFATC1
Genomic Map