Assay Details
| Gene Symbol: PAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095275.4 | XP_001095275.3 | 918 | 87 | ||
| XM_015149591.2 | XP_015005077.1 | 918 | 87 | |||
| GenBank mRNA | JU487536.1 | - | 770 | 87 | ||
More Information
Set Membership: |
|

PAX1
Genomic Map