Assay Details
| Gene Symbol: PLCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260854.1 | NP_001247783.1 | 3411 | 99 | ||
| XM_028830422.1 | XP_028686255.1 | 2989 | 99 | |||
| GenBank mRNA | JU321317.1 | - | 3184 | 99 | ||
| JU321318.1 | - | 3184 | 99 | |||
| JU476025.1 | - | 3411 | 99 | |||
More Information
Set Membership: |
|

PLCL1
Genomic Map