Assay Details
| Gene Symbol: CDIPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260643.1 | NP_001247572.1 | 719 | 85 | ||
| XM_015125953.2 | XP_014981439.1 | 1139 | 85 | |||
| XM_028840493.1 | XP_028696326.1 | 1199 | 85 | |||
| GenBank mRNA | JU321684.1 | - | 723 | 85 | ||
| JU472849.1 | - | 712 | 85 | |||
| JV044080.1 | - | 719 | 85 | |||
| JV567513.1 | - | 727 | 85 | |||
More Information
Set Membership: |
|

CDIPT
Genomic Map