Assay Details
| Gene Symbol: TIMM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190973.2 | NP_001177902.1 | 386 | 79 | ||
| XM_028853456.1 | XP_028709289.1 | 371 | 79 | |||
| GenBank mRNA | JU320757.1 | - | 385 | 79 | ||
| JU471088.1 | - | 391 | 79 | |||
| JV044525.1 | - | 389 | 79 | |||
| JV491241.1 | - | 305 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028853456.1 transcripts.
Set Membership: |
|

TIMM23
Genomic Map