Assay Details
| Gene Symbol: LOC106992427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113620.2 | XP_014969106.1 | 731 | 63 | ||
| GenBank mRNA | JU319931.1 | - | 420 | 63 | ||
| JU473981.1 | - | 442 | 63 | |||
| JV045906.1 | - | 452 | 63 | |||
| JV640194.1 | - | 485 | 63 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC106992427
Genomic Map