Assay Details
| Gene Symbol: TIMM17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195660.1 | NP_001182589.1 | 151 | 131 | ||
| GenBank mRNA | CB308818.1 | - | 154 | 131 | ||
| CO644659.1 | - | 137 | 131 | |||
| JU472778.1 | - | 139 | 131 | |||
| JV045909.1 | - | 155 | 131 | |||
| JV635773.1 | - | 163 | 131 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMM17A
Genomic Map