Assay Details
| Gene Symbol: CRTAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261752.1 | NP_001248681.1 | 878 | 91 | ||
| GenBank mRNA | JU332521.1 | - | 888 | 91 | ||
| JU473603.1 | - | 848 | 91 | |||
| JV045924.1 | - | 878 | 91 | |||
More Information
Set Membership: |
|

CRTAP
Genomic Map