Assay Details
| Gene Symbol: RPP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087609.4 | XP_001087609.1 | 322 | 64 | ||
| XM_015147668.2 | XP_015003154.1 | 322 | 64 | |||
| GenBank mRNA | JU478902.1 | - | 285 | 64 | ||
| JV640339.1 | - | 297 | 64 | |||
More Information
Set Membership: |
|

RPP30
Genomic Map