Assay Details
| Gene Symbol: RABAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260923.1 | NP_001247852.1 | 536 | 53 | ||
| GenBank mRNA | JU322608.1 | - | 547 | 53 | ||
| JV045180.1 | - | 536 | 53 | |||
| JV490048.1 | - | 528 | 53 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RABAC1
Genomic Map