Assay Details
| Gene Symbol: LOC717978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120768.2 | XP_014976254.2 | 4833 | 81 | ||
| GenBank mRNA | JU333035.1 | - | 4830 | 81 | ||
| JU333036.1 | - | 4830 | 81 | |||
| Gene Symbol: PARP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837587.1 | XP_028693420.1 | 4826 | 81 | ||
| GenBank mRNA | JV046242.1 | - | 4798 | 81 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC717978
Genomic Map