Assay Details
| Gene Symbol: MXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118504.4 | XP_001118504.3 | 614 | 70 | ||
| XM_028848237.1 | XP_028704070.1 | 614 | 70 | |||
| GenBank mRNA | JU321168.1 | - | 343 | 70 | ||
| JV043853.1 | - | 337 | 70 | |||
| JV527869.1 | - | 344 | 70 | |||
More Information
Set Membership: |
|

MXD4
Genomic Map