Assay Details
| Gene Symbol: CLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121224.2 | XP_014976710.1 | 688 | 85 | ||
| GenBank mRNA | JU325709.1 | - | 234 | 85 | ||
| JV605492.1 | - | 275 | 85 | |||
| JV706580.1 | - | 236 | 85 | |||
More Information
Set Membership: |
|

CLN5
Genomic Map