Assay Details
| Gene Symbol: ERF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124066.2 | XP_014979552.1 | 545 | 112 | ||
| XM_028838884.1 | XP_028694717.1 | 402 | 112 | |||
| GenBank mRNA | JU474654.1 | - | 494 | 112 | ||
| JV043658.1 | - | 507 | 112 | |||
| JV574251.1 | - | 517 | 112 | |||
More Information
Set Membership: |
|

ERF
Genomic Map