Assay Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114509.4 | XP_001114509.1 | 1928 | 126 | ||
| XM_015110852.2 | XP_014966338.1 | 1967 | 126 | |||
| XM_028828554.1 | XP_028684387.1 | 466 | 126 | |||
| XM_028828555.1 | XP_028684388.1 | 211 | 126 | |||
| GenBank mRNA | JU322560.1 | - | 724 | 126 | ||
| JU473346.1 | - | 724 | 126 | |||
| JV043921.1 | - | 739 | 126 | |||
| JV639214.1 | - | 754 | 126 | |||
| JV704382.1 | - | 717 | 126 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PMVK
Genomic Map