Assay Details
| Gene Symbol: DCTN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257422.1 | NP_001244351.1 | 147 | 127 | ||
| XM_028852117.1 | XP_028707950.1 | 463 | 127 | |||
| GenBank mRNA | JU320748.1 | - | 147 | 127 | ||
| JU474042.1 | - | 151 | 127 | |||
| JV045964.1 | - | 151 | 127 | |||
| JV601650.1 | - | 180 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DCTN6
Genomic Map