Assay Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113075.2 | XP_014968561.1 | 1088 | 152 | ||
| XM_028832278.1 | XP_028688111.1 | 1088 | 152 | |||
| XM_028832280.1 | XP_028688113.1 | 1358 | 152 | |||
| GenBank mRNA | JU341876.1 | - | 839 | 152 | ||
| JV648453.1 | - | 879 | 152 | |||
More Information
Set Membership: |
|

SULT1C4
Genomic Map