Assay Details
| Gene Symbol: PPP1R17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193720.1 | NP_001180649.1 | 706 | 66 | ||
| XM_028844786.1 | XP_028700619.1 | 156 | 66 | |||
| GenBank mRNA | JU322484.1 | - | 202 | 66 | ||
More Information
Set Membership: |
|

PPP1R17
Genomic Map