Assay Details
| Gene Symbol: GADD45G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193910.1 | NP_001180839.1 | 471 | 95 | ||
| GenBank mRNA | JV043793.1 | - | 474 | 95 | ||
| JV638916.1 | - | 471 | 95 | |||
More Information
Set Membership: |
|

GADD45G
Genomic Map