Assay Details
| Gene Symbol: GLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261764.1 | NP_001248693.1 | 562 | 73 | ||
| XM_015136263.2 | XP_014991749.1 | 636 | 73 | |||
| GenBank mRNA | JU322612.1 | - | 575 | 73 | ||
| JU473595.1 | - | 542 | 73 | |||
| JV045135.1 | - | 562 | 73 | |||
| JV465098.1 | - | 430 | 73 | |||
| JV602384.1 | - | 579 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GLO1
Genomic Map