Assay Details
| Gene Symbol: DBF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134146.2 | XP_014989632.1 | 1032 | 75 | ||
| XM_015134147.2 | XP_014989633.1 | 854 | 75 | |||
| GenBank mRNA | JU474089.1 | - | 1046 | 75 | ||
| JV705673.1 | - | 1030 | 75 | |||
More Information
Set Membership: |
|

DBF4
Genomic Map