Assay Details
| Gene Symbol: SIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113772.2 | XP_014969258.1 | 857 | 146 | ||
| XM_015113773.2 | XP_014969259.1 | 1419 | 146 | |||
| XM_015113774.2 | XP_014969260.1 | 1004 | 146 | |||
| XM_028833230.1 | XP_028689063.1 | 857 | 146 | |||
| GenBank mRNA | JU324245.1 | - | 822 | 146 | ||
| JU476849.1 | - | 841 | 146 | |||
More Information
Set Membership: |
|

SIPA1
Genomic Map